1-(5-Nitropyridin-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A611374-100MG A611374-1G A611374-250MG
76831-010EA 57.29 USD
76831-010 76831-012 76831-014
1-(5-Nitropyridin-2-yl)ethanone 95%
1-(5-Nitropyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆N₂O₃
MW: 166.13 g/mol
MDL Number: MFCD10697712
CAS Number: 31557-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR