4,8-Dibromoquinoline 97%

Supplier: Ambeed
Warning

A610993-10G A610993-1G A610993-250MG A610993-5G
76831-000EA 241 USD
76831-000 76831-002 76831-004 77613-902
4,8-Dibromoquinoline 97%
4,8-Dibromoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Br₂N
MDL Number: MFCD08063207
CAS Number: 1070879-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR