3-Fluoro-1,1'-biphenyl 95%

Supplier: Ambeed
Warning

A609091-1G A609091-5G
76830-996EA 31.83 USD
76830-996 76830-998
3-Fluoro-1,1'-biphenyl 95%
3-Fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₉F
MW: 172.20 g/mol
MDL Number: MFCD01740506
CAS Number: 2367-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR