6-(Trifluoromethyl)-4,5-dihydropyridazin-3(2H)-one 95+%

Supplier: Ambeed
Warning

A608399-1G A608399-5G
76830-994EA 45 USD
76830-994 77886-458
6-(Trifluoromethyl)-4,5-dihydropyridazin-3(2H)-one 95+%
6-(Trifluoromethyl)-4,5-dihydropyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂O
MDL Number: MFCD13705400
CAS Number: 628332-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR