Methyl 4-amino-1H-indazole-6-carboxylate 97%

Supplier: Ambeed
Danger

A608165-1G A608165-250MG
76830-990EA 116.7 USD
76830-990 76830-992
Methyl 4-amino-1H-indazole-6-carboxylate 97%
Methyl 4-amino-1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MW: 191.18 g/mol
MDL Number: MFCD07781320
CAS Number: 885518-51-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR