2-Bromo-1-(3,5-dichlorophenyl)ethanone 97%

Supplier: Ambeed
Danger

A607090-10G A607090-250MG
76830-980EA 279.02 USD
76830-980 76830-982
2-Bromo-1-(3,5-dichlorophenyl)ethanone 97%
2-Bromo-1-(3,5-dichlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O
MDL Number: MFCD03425180
CAS Number: 53631-13-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR