3-Bromo-2-iodoaniline 97%

Supplier: Ambeed
Warning

A607005-100MG A607005-1G A607005-250MG A607005-5G
76830-974EA 47 USD
76830-974 76830-976 76830-978 77886-454
3-Bromo-2-iodoaniline 97%
3-Bromo-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅BrIN
MDL Number: MFCD16622211
CAS Number: 150908-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR