(6-Phenoxypyridin-3-yl)boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 2-Phenoxypyridine-5-boronic acid

A605988-100MG A605988-1G A605988-250MG
76830-954EA 37 USD
76830-954 76830-956 76830-958
(6-Phenoxypyridin-3-yl)boronic acid 96%
(6-Phenoxypyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₃
MW: 215.01 g/mol
MDL Number: MFCD12403506
CAS Number: 1270921-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR