Thiomorpholine 98%, Ambeed.Inc

Supplier: Ambeed
Danger

A603893-1G A603893-25G A603893-5G A603893-10G A603893-100G
76830-946EA 5.31 USD
76830-946 76830-948 76830-950 77551-816 77684-474
Thiomorpholine 98%, Ambeed.Inc
Thiomorpholine

Precaución: For research use only.

Formula: C₄H₉NS
MW: 103.19 g/mol
MDL Number: MFCD00005974
CAS Number: 123-90-0
UN: 3267
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR