3-Amino-1H-pyrazolo[3,4-c]pyridine 98%

Supplier: Ambeed
Danger

Synonyms: 1H-Pyrazolo[3,4-c]pyridin-3-amine, 3-Amino-6-azaindole

A603448-1G A603448-250MG A603448-5G
76830-940EA 224.92 USD
76830-940 76830-942 76830-944
3-Amino-1H-pyrazolo[3,4-c]pyridine 98%
3-Amino-1H-pyrazolo[3,4-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₆N₄
MW: 134.139 g/mol
MDL Number: MFCD11656373
CAS Number: 76006-17-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR