3-Methyl-2-naphthol 98%

Supplier: Ambeed
Danger

Synonyms: 2-Hydroxy-3-methylnaphthalene, 3-Methylnaphthalen-2-ol

A602020-100MG A602020-10G A602020-1G A602020-250MG A602020-5G
76830-930EA 28.65 USD
76830-930 76830-932 76830-934 76830-936 76830-938
3-Methyl-2-naphthol 98%
3-Methyl-2-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.1965 g/mol
MDL Number: MFCD09037754
CAS Number: 17324-04-8
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR