1-Bromo-4-(methoxymethoxy)benzene 98%

Supplier: Ambeed
Warning

A601388-1G A601388-250MG A601388-25G A601388-5G
76830-924EA 49.87 USD
76830-924 76830-926 76830-928 77572-944
1-Bromo-4-(methoxymethoxy)benzene 98%
1-Bromo-4-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.0599 g/mol
MDL Number: MFCD00457268
CAS Number: 25458-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR