5-Bromo-3-fluoro-2-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A600817-100MG A600817-1G A600817-250MG
76830-918EA 13 USD
76830-918 76830-920 76830-922
5-Bromo-3-fluoro-2-hydroxybenzoic acid 95%
5-Bromo-3-fluoro-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrFO₃
MDL Number: MFCD06203615
CAS Number: 251300-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR