2-Bromonaphthalene-1,4-dione 97%

Supplier: Ambeed
Warning

A599793-1G A599793-5G
76830-914EA 16 USD
76830-914 76830-916
2-Bromonaphthalene-1,4-dione 97%
2-Bromonaphthalene-1,4-dione

Precaución: for Research Use Only

Formula: C₁₀H₅BrO₂
MW: 237.04 g/mol
MDL Number: MFCD00629301
CAS Number: 2065-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR