(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate 98%

Supplier: Ambeed
Warning

Synonyms: (11bS)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

A599460-100MG A599460-1G
76830-910EA 35 USD
76830-910 76830-912
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate 98%
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₃₆H₁₇F₁₂O₄P
MDL Number: MFCD08689863
CAS Number: 878111-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR