Methyl 3-oxoisoindoline-5-carboxylate 97%

Supplier: Ambeed
Warning

A599456-100MG A599456-250MG
76830-908EA 22 USD
76830-908 77886-451
Methyl 3-oxoisoindoline-5-carboxylate 97%
Methyl 3-oxoisoindoline-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MDL Number: MFCD09878911
CAS Number: 954239-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR