3-Bromo-6-methyl-5-nitropyridin-2-ol 98%

Supplier: Ambeed
Warning

A599022-1G A599022-250MG A599022-25G A599022-5G
76830-902EA 45.62 USD
76830-902 76830-904 76830-906 77684-470
3-Bromo-6-methyl-5-nitropyridin-2-ol 98%
3-Bromo-6-methyl-5-nitropyridin-2-ol

Precaución: For research use only.

Formula: C₆H₅BrN₂O₃
MW: 233.01 g/mol
MDL Number: MFCD09839283
CAS Number: 874493-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR