(R)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A598013-100MG A598013-250MG A598013-5G
76830-886EA 14.86 USD
76830-886 76830-888 77684-468
(R)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene 98%, Ambeed.Inc
(R)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene

Precaución: For research use only.

Formula: C₂₄H₂₀I₂O₄
MW: 626.22 g/mol
CAS Number: 189518-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR