6-Hydroxy-pyrrolo[3,4-b]pyridine-5,7-dione 95+%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxy-5H-pyrrolo[3,4-b]pyridine-5,7(6H)-dione

A595876-1G A595876-250MG
76830-874EA 210.06 USD
76830-874 76830-876
6-Hydroxy-pyrrolo[3,4-b]pyridine-5,7-dione 95+%
6-Hydroxy-pyrrolo[3,4-b]pyridine-5,7-dione

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
MW: 164.118 g/mol
MDL Number: MFCD11865269
CAS Number: 23439-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR