Methoxy(cyclooctadiene)rhodium(I) dimer ≥98% Rh> 42%

Supplier: Ambeed
Warning

A593725-50MG A593725-100MG A593725-250MG
76830-858EA 93 USD
76830-858 77886-436 77886-437
Methoxy(cyclooctadiene)rhodium(I) dimer ≥98% Rh> 42%
Methoxy(cyclooctadiene)rhodium(I) dimer

Precaución: for Research Use Only

Formula: C₁₈H₃₀O₂Rh₂
MDL Number: MFCD08457639
CAS Number: 12148-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR