4-Bromo-7-methoxy-1H-indazole 98%

Supplier: Ambeed
Warning

A593140-100MG A593140-1G A593140-250MG
76830-852EA 34 USD
76830-852 76830-854 76830-856
4-Bromo-7-methoxy-1H-indazole 98%
4-Bromo-7-methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MDL Number: MFCD17016112
CAS Number: 938062-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR