(4-(Propylsulfonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A591186-1G A591186-250MG A591186-5G
76830-846EA 160 USD
76830-846 76830-848 76830-850
(4-(Propylsulfonyl)phenyl)boronic acid 98%
(4-(Propylsulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₄S
MW: 228.07 g/mol
MDL Number: MFCD13195655
CAS Number: 1217501-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR