Methyl 7-bromoquinoline-4-carboxylate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A590514-250MG A590514-1G A590514-100MG
76830-842EA 20 USD
76830-842 77684-455 77886-430
Methyl 7-bromoquinoline-4-carboxylate 95%, Ambeed.Inc
Methyl 7-bromoquinoline-4-carboxylate

Precaución: For research use only.

Formula: C₁₁H₈BrNO₂
MDL Number: MFCD11976157
CAS Number: 220844-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR