tert-Butyl (chloromethyl) succinate 95%

Supplier: Ambeed
Danger

A588696-100MG A588696-250MG A588696-5G A588696-1G
76830-832EA 19 USD
76830-832 76830-834 77594-790 77886-429
tert-Butyl (chloromethyl) succinate 95%
tert-Butyl (chloromethyl) succinate

Precaución: for Research Use Only

Formula: C₉H₁₅ClO₄
MW: 222.66 g/mol
MDL Number: MFCD13194872
CAS Number: 432037-43-9
UN: 3265
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR