1-Octanesulfonic acid sodium salt monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate

A588428-5G A588428-500G
76830-830EA 12.74 USD
76830-830 77684-453
1-Octanesulfonic acid sodium salt monohydrate 98%
1-Octanesulphonic acid sodium salt monohydrate

Precaución: For research use only.

Formula: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
MDL Number: MFCD00007544
CAS Number: 207596-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR