1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide 98%

Supplier: Ambeed
Warning

Synonyms: PYR14-TFSI, N-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulphonyl)imide

A587758-100G A587758-5G A587758-1G A587758-25G
76830-828EA 158 USD
76830-828 77572-872 77594-786 77886-426
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide 98%
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide

Precaución: for Research Use Only

Formula: C₁₁H₂₀F₆N₂O₄S₂
MW: 422.41
MDL Number: MFCD07784447
CAS Number: 223437-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR