8-Bromo-6-chloroimidazo[1,2-a]pyridine-3-carbaldehyde 96%

Supplier: Ambeed
Warning

A585924-100MG A585924-1G A585924-250MG
76830-812EA 5 USD
76830-812 76830-814 76830-816
8-Bromo-6-chloroimidazo[1,2-a]pyridine-3-carbaldehyde 96%
8-Bromo-6-chloroimidazo[1,2-a]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂O
MDL Number: MFCD11504922
CAS Number: 1093092-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR