5-(3-Nitrophenyl)isoxazole-3-carboxylic acid ethyl ester 97+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-(3-nitrophenyl)isoxazole-3-carboxylate

A585501-1G A585501-250MG A585501-5G
76830-806EA 44 USD
76830-806 76830-808 76830-810
5-(3-Nitrophenyl)isoxazole-3-carboxylic acid ethyl ester 97+%
5-(3-Nitrophenyl)isoxazole-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₅
MDL Number: MFCD03848619
CAS Number: 866040-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR