Fmoc-His(Fmoc)-OH 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: N,1-Di-Fmoc-L-histidine

A585258-1G A585258-25G A585258-5G
76830-800EA 8 USD
76830-800 76830-802 76830-804
Fmoc-His(Fmoc)-OH 98% (by HPLC)
Fmoc-His(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₃₆H₂₉N₃O₆
MW: 599.643 g/mol
MDL Number: MFCD00070658
CAS Number: 98929-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR