2-Chloro-3,6-dimethylpyridine 95%

Supplier: Ambeed
Warning

A582848-1G A582848-100MG A582848-250MG
76830-792EA 137 USD
76830-792 77886-415 77886-416
2-Chloro-3,6-dimethylpyridine 95%
2-Chloro-3,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MDL Number: MFCD12400985
CAS Number: 72093-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR