2-(2-(2-Hydroxyethoxy)ethyl)isoindoline-1,3-dione 97%

Supplier: Ambeed
Warning

A582223-250MG A582223-5G A582223-25G
76830-790EA 31.83 USD
76830-790 77613-804 77684-438
2-(2-(2-Hydroxyethoxy)ethyl)isoindoline-1,3-dione 97%
2-(2-(2-Hydroxyethoxy)ethyl)isoindoline-1,3-dione

Precaución: For research use only.

Formula: C₁₂H₁₃NO₄
MW: 235.23 g/mol
MDL Number: MFCD06661363
CAS Number: 69676-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR