4-Bromo-3-methylthiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A580822-1G A580822-250MG A580822-5G
76830-774EA 38.2 USD
76830-774 76830-776 77594-764
4-Bromo-3-methylthiophene-2-carboxylic acid 95%
4-Bromo-3-methylthiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅BrO₂S
MW: 221.07 g/mol
MDL Number: MFCD03789231
CAS Number: 265652-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR