2,2-Dimethyl-5-(propan-2-ylidene)-1,3-dioxane-4,6-dione 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A580069-100MG A580069-250MG A580069-5G A580069-1G
76830-768EA 12 USD
76830-768 76830-770 76830-772 77684-432
2,2-Dimethyl-5-(propan-2-ylidene)-1,3-dioxane-4,6-dione 98%, Ambeed.Inc
2,2-Dimethyl-5-(propan-2-ylidene)-1,3-dioxane-4,6-dione

Precaución: For research use only.

Formula: C₉H₁₂O₄
MDL Number: MFCD16038481
CAS Number: 2231-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR