(2-Formyl-4,5-dimethoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A578615-1G A578615-250MG A578615-5G
76830-758EA 62.6 USD
76830-758 76830-760 76830-762
(2-Formyl-4,5-dimethoxyphenyl)boronic acid 98%
(2-Formyl-4,5-dimethoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MW: 209.99 g/mol
MDL Number: MFCD16660278
CAS Number: 1005346-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR