5-Methoxy-2,4-dimethylpyrimidine 95%

Supplier: Ambeed
Warning

A578017-1G A578017-250MG
76830-754EA 380.87 USD
76830-754 76830-756
5-Methoxy-2,4-dimethylpyrimidine 95%
5-Methoxy-2,4-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD23135769
CAS Number: 1369766-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR