6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol 95%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxynaphthalene-2-boronic acid pinacol ester

A576776-1G A576776-250MG A576776-5G
76830-744EA 19 USD
76830-744 76830-746 76830-748
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol 95%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₉BO₃
MW: 270.135 g/mol
MDL Number: MFCD20486767
CAS Number: 269410-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR