Ethyl 4-(hydroxymethyl)picolinate 95%

Supplier: Ambeed
Warning

A576111-100MG A576111-250MG
76830-740EA 23 USD
76830-740 76830-742
Ethyl 4-(hydroxymethyl)picolinate 95%
Ethyl 4-(hydroxymethyl)picolinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD09844039
CAS Number: 59663-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR