2-Chloro-5-iodonicotinonitrile 98%

Supplier: Ambeed
Warning

A574264-1G A574264-250MG A574264-5G
76830-732EA 57.29 USD
76830-732 76830-734 76830-736
2-Chloro-5-iodonicotinonitrile 98%
2-Chloro-5-iodonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂ClIN₂
MW: 264.451 g/mol
MDL Number: MFCD09909360
CAS Number: 766515-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR