2-(Furan-2-yl)-7-(3-(4-methoxyphenyl)propyl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine 98%

Supplier: Ambeed
Warning

Synonyms: SCH442416

A573317-10MG A573317-1MG A573317-25MG
76830-726EA 103.97 USD
76830-726 76830-728 76830-730
2-(Furan-2-yl)-7-(3-(4-methoxyphenyl)propyl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine 98%
2-(Furan-2-yl)-7-(3-(4-methoxyphenyl)propyl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₂₀H₁₉N₇O₂
MW: 389.41 g/mol
MDL Number: MFCD08703126
CAS Number: 316173-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR