Methyl 5-bromo-2-methylthiophene-3-carboxylate 95%

Supplier: Ambeed
Warning

A573104-100MG A573104-1G
76830-722EA 114.58 USD
76830-722 76830-724
Methyl 5-bromo-2-methylthiophene-3-carboxylate 95%
Methyl 5-bromo-2-methylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
MW: 235.09 g/mol
MDL Number: MFCD14636529
CAS Number: 1259396-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR