2,6-Dimethyl-4-nitropyridine 97%

Supplier: Ambeed
Warning

A568869-1G A568869-250MG
76830-704EA 114.58 USD
76830-704 76830-706
2,6-Dimethyl-4-nitropyridine 97%
2,6-Dimethyl-4-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.151 g/mol
MDL Number: MFCD00234324
CAS Number: 4913-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR