1-Methyl-5-(methylsulfanyl)-1H-1,2,4-triazol-3-ylamine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-1-methyl-5-methylthio-1,2,4-triazole

A568596-1G A568596-250MG
76830-700EA 68.96 USD
76830-700 76830-702
1-Methyl-5-(methylsulfanyl)-1H-1,2,4-triazol-3-ylamine 95%
1-Methyl-5-(methylsulfanyl)-1H-1,2,4-triazol-3-ylamine

Precaución: for Research Use Only

Formula: C₄H₈N₄S
MW: 144.2 g/mol
MDL Number: MFCD00051817
CAS Number: 84827-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR