2-Iodo-4-methoxybenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A566679-100MG
A566679-1G
A566679-250MG
A566679-5G
A566679-10G
76830-676EA
7.43
USD
76830-676
76830-678
76830-680
76830-682
77613-772
2-Iodo-4-methoxybenzoic acid 95%
2-Iodo-4-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇IO₃ |
MDL Number:
MFCD09909619 CAS Number: 54435-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...