Spiro[2.2]pentane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

A566593-100MG A566593-1G A566593-250MG A566593-50MG A566593-5G
76830-666EA 126.01 USD
76830-666 76830-668 76830-670 76830-672 76830-674
Spiro[2.2]pentane-1-carboxylic acid 95%
Spiro[2.2]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₂
MDL Number: MFCD04038995
CAS Number: 17202-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR