5-Fluoro-1H-pyrrolo[2,3-b]pyridin-6-amine 97%

Supplier: Ambeed
Warning

A566307-100MG A566307-1G A566307-250MG
76830-660EA 97.61 USD
76830-660 76830-662 76830-664
5-Fluoro-1H-pyrrolo[2,3-b]pyridin-6-amine 97%
5-Fluoro-1H-pyrrolo[2,3-b]pyridin-6-amine

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MW: 151.14 g/mol
MDL Number: MFCD12962828
CAS Number: 1190321-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR