3-(Cyclopentyloxy)-5-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

A564559-100MG A564559-1G A564559-250MG
76830-654EA 39 USD
76830-654 76830-656 76830-658
3-(Cyclopentyloxy)-5-methylphenylboronic acid 98%
3-(Cyclopentyloxy)-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₃
MDL Number: MFCD16295068
CAS Number: 1256346-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR