4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2,6,6-Tetramethyl-4-(benzoyloxy)piperidine-1-oxyl, 4-Benzoyloxy-TEMPO, 4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Hydroxy-TEMPO Benzoate
A564526-1G
A564526-250MG
A564526-25G
A564526-5G
76830-646EA
24.41
USD
76830-646
76830-648
76830-650
76830-652
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate 98%
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate
Precaución: for Research Use Only
Formula:
C₁₆H₂₂NO₃ MW: 276.36 g/mol |
MDL Number:
MFCD00075563 CAS Number: 3225-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...