4-Amino-3-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A563747-100MG A563747-1G A563747-250MG A563747-5G
76830-640EA 23 USD
76830-640 76830-642 76830-644 77886-376
4-Amino-3-nitrobenzaldehyde 97%
4-Amino-3-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆N₂O₃
MDL Number: MFCD17170817
CAS Number: 51818-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR