2-Oxa-6-azaspiro[3.4]octan-7-one 95%

Supplier: Ambeed
Warning

A562646-100MG A562646-1G A562646-250MG
76830-628EA 94.43 USD
76830-628 76830-630 76830-632
2-Oxa-6-azaspiro[3.4]octan-7-one 95%
2-Oxa-6-azaspiro[3.4]octan-7-one

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MDL Number: MFCD14586414
CAS Number: 1207174-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR