2-Butoxy-5-chlorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Butoxy-5-chlorophenyl)boronic acid

A558212-100MG A558212-1G A558212-250MG
76830-616EA 12 USD
76830-616 76830-618 76830-620
2-Butoxy-5-chlorophenylboronic acid 98%
2-Butoxy-5-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClO₃
MDL Number: MFCD08276746
CAS Number: 352534-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR